Merck
Influence of green tea extract on oxidative damage and apoptosis induced by deltamethrin in rat brain.
Neurotoxicology and teratology (2015-05-28)
Hanan A Ogaly, A A Khalaf, Marwa A Ibrahim, Mona K Galal, Reham M Abd-Elsalam
ABSTRAKT
In the present study, we investigated the protective effect of an aqueous extract of green tea leaves (GTE) against neurotoxicity and oxidative damage induced by deltamethrin (DM) in male rats. Four different groups of rats were used: the 1st group was the vehicle treated control group, the 2nd group received DM (0.6 mg/kg BW), the 3rd group received DM plus GTE, and the 4th received GTE alone (25 mg/kg BW). The brain tissues were collected at the end of the experimental regimen for subsequent investigation. Rats that were given DM had a highly significant elevation in MDA content, nitric oxide concentration, DNA fragmentation and expression level of apoptotic genes, TP53 and COX2. Additionally, a significant reduction in the total antioxidant capacity in the second group was detected. The findings for the 3rd group highlight the efficacy of GTE as a neuro-protectant in DM-induced neurotoxicity through improving the oxidative status and DNA fragmentation as well as suppressing the expression of the TP53 and COX2 genes. In conclusion, GTE, at a concentration of 25mg/kg/day, protected against DM-induced neurotoxicity through its antioxidant and antiapoptotic influence; therefore, it can be used as a protective natural product against DM-induced neurotoxicity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
3,3′-Diaminobenzidine tetrahydrochloride, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer