Merck
Angiotensin-converting enzyme insertion/deletion gene polymorphisms is associated with risk of glioma in a Chinese population.
Journal of the renin-angiotensin-aldosterone system : JRAAS (2014-08-22)
MinXue Lian, HaiTao Jiang, Hui Wang, ShiWen Guo
ABSTRAKT
The insertion/deletion (I/D) polymorphism of the angiotensin-converting enzyme (ACE) gene has recently been linked to the pathogenesis and progression of human cancers. The aim of this study was to evaluate the potential association between ACE I/D polymorphism and glioma in a Chinese population. A case-control study involving patients with 800 glioma and 800 controls was conducted. Polymerase chain reaction restriction fragment length polymorphism (PCR-RFLP) assay was applied to assess the ACE I/D genotypes. Glioma cases had a significantly higher frequency of DD genotype [odds ratio (OR) = 1.61, 95% confidence interval (CI) = 1.12, 2.32; p = 0.01] than controls. When stratified by the grade of glioma, cases with WHO IV glioma had a significantly higher frequency of DD genotype (OR = 1.51, 95% CI = 1.03, 2.21; p = 0.03). When stratified by the histology of glioma, there was no significant difference in the distribution of each genotype. Our study suggested that the ACE DD genotype was associated with a higher glioma risk in this Chinese population. To the best of our knowledge, this is the first report describing the potential association between ACE I/D polymorphism and glioma. Additional studies are needed to confirm this finding.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder