Merck
A highly selective biosynthetic pathway to non-natural C50 carotenoids assembled from moderately selective enzymes.
Nature communications (2015-07-15)
Maiko Furubayashi, Mayu Ikezumi, Shinichi Takaichi, Takashi Maoka, Hisashi Hemmi, Takuya Ogawa, Kyoichi Saito, Alexander V Tobias, Daisuke Umeno
ABSTRAKT
Synthetic biology aspires to construct natural and non-natural pathways to useful compounds. However, pathways that rely on multiple promiscuous enzymes may branch, which might preclude selective production of the target compound. Here, we describe the assembly of a six-enzyme pathway in Escherichia coli for the synthesis of C50-astaxanthin, a non-natural purple carotenoid. We show that by judicious matching of engineered size-selectivity variants of the first two enzymes in the pathway, farnesyl diphosphate synthase (FDS) and carotenoid synthase (CrtM), branching and the production of non-target compounds can be suppressed, enriching the proportion of C50 backbones produced. We then further extend the C50 pathway using evolved or wild-type downstream enzymes. Despite not containing any substrate- or product-specific enzymes, the resulting pathway detectably produces only C50 carotenoids, including ∼ 90% C50-astaxanthin. Using this approach, highly selective pathways can be engineered without developing absolutely specific enzymes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
L-(+)-Arabinose, 99%
Sigma-Aldrich
L-(+)-Arabinose, ≥99% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
L-(+)-Arabinose, 98%
Sigma-Aldrich
Carbenicillin, Ready Made Solution, 100 mg/mL in ethanol/water
Sigma-Aldrich
Chloramphenicol, ≥98% (HPLC)
Sigma-Aldrich
Chloramphenicol, meets USP testing specifications
Sigma-Aldrich
Chloramphenicol, BioReagent, suitable for plant cell culture
Sigma-Aldrich
3-Nitrobenzyl alcohol, 98%