Merck
  • Elevated glycated albumin and reduced endogenous secretory receptor for advanced glycation endproducts levels in serum predict major adverse cardio-cerebral events in patients with type 2 diabetes and stable coronary artery disease.
Elevated glycated albumin and reduced endogenous secretory receptor for advanced glycation endproducts levels in serum predict major adverse cardio-cerebral events in patients with type 2 diabetes and stable coronary artery disease.
International journal of cardiology (2015-07-06)
Zhen Kun Yang, Ying Shen, Wei Feng Shen, Li Jin Pu, Hua Meng, Rui Yan Zhang, Qi Zhang, Qiu Jing Chen, Raffaele De Caterina, Lin Lu
ABSTRAKT
Glycated albumin (GA) and the endogenous secretory receptor for advanced glycation endproducts (esRAGE) may modulate risk related to atherosclerosis. We tested the hypothesis that elevated GA and reduced esRAGE in serum are associated with adverse clinical outcomes in patients with type 2 diabetes and stable coronary artery disease (CAD). We determined GA and esRAGE serum levels in 576 consecutive patients with type 2 diabetes and stable CAD undergoing sirolimus-eluting stent (SES)-PCI. The primary endpoint was the incidence of major adverse cardio-cerebral events (MACCE) including cardiac death, non-fatal myocardial infarction, and non-fatal stroke during a 2-year follow-up. The secondary endpoint was the occurrence of clinically driven repeat revascularization during a 2-year follow-up. The prognostic value of GA and esRAGE was determined with the Cox-proportional hazard model after adjustment for covariates. A total 40 patients (6.9%) experienced MACCE, and 108 (18.8%) patients underwent repeat coronary revascularization during the follow-up. Serum GA (HR=1.22, 95% CI 1.16-1.28; HR=1.15, 95% CI 1.11-1.19, respectively; for both p<0.001) and esRAGE (HR=0.60, 95% CI 0.40-0.87; HR=0.75, 95% CI 0.61-0.92, respectively; for both p<0.01) levels remained independent predictors of the primary and secondary endpoints after adjustment for possible confounders. Serum GA and esRAGE are novel predictors of long-term clinical outcomes in patients with type 2 diabetes and stable CAD. Increased serum GA and decreased esRAGE are associated with a poor prognosis in such patients.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder