Merck
Imaging metabolite dynamics in living cells using a Spinach-based riboswitch.
Proceedings of the National Academy of Sciences of the United States of America (2015-05-13)
Mingxu You, Jacob L Litke, Samie R Jaffrey
ABSTRAKT
Riboswitches are natural ligand-sensing RNAs typically that are found in the 5' UTRs of mRNA. Numerous classes of riboswitches have been discovered, enabling mRNA to be regulated by diverse and physiologically important cellular metabolites and small molecules. Here we describe Spinach riboswitches, a new class of genetically encoded metabolite sensor derived from naturally occurring riboswitches. Drawing upon the structural switching mechanism of natural riboswitches, we show that Spinach can be swapped for the expression platform of various riboswitches, allowing metabolite binding to induce Spinach fluorescence directly. In the case of the thiamine 5'-pyrophosphate (TPP) riboswitch from the Escherichia coli thiM gene encoding hydroxyethylthiazole kinase, we show that insertion of Spinach results in an RNA sensor that exhibits fluorescence upon binding TPP. This TPP Spinach riboswitch binds TPP with affinity and selectivity similar to that of the endogenous riboswitch and enables the discovery of agonists and antagonists of the TPP riboswitch using simple fluorescence readouts. Furthermore, expression of the TPP Spinach riboswitch in Escherichia coli enables live imaging of dynamic changes in intracellular TPP concentrations in individual cells. Additionally, we show that other riboswitches that use a structural mechanism similar to that of the TPP riboswitch, including the guanine and adenine riboswitches from the Bacillus subtilis xpt gene encoding xanthine phosphoribosyltransferase, and the S-adenosyl-methionine-I riboswitch from the B. subtilis yitJ gene encoding methionine synthase, can be converted into Spinach riboswitches. Thus, Spinach riboswitches constitute a novel class of RNA-based fluorescent metabolite sensors that exploit the diversity of naturally occurring ligand-binding riboswitches.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
S-(5′-Adenosyl)-L-homocysteine, crystalline