Merck
Developmental exposure to bisphenol A (BPA) alters sexual differentiation in painted turtles (Chrysemys picta).
General and comparative endocrinology (2015-04-12)
Caitlin M Jandegian, Sharon L Deem, Ramji K Bhandari, Casey M Holliday, Diane Nicks, Cheryl S Rosenfeld, Kyle W Selcer, Donald E Tillitt, Frederick S Vom Saal, Vanessa Vélez-Rivera, Ying Yang, Dawn K Holliday
ABSTRAKT
Environmental chemicals can disrupt endocrine signaling and adversely impact sexual differentiation in wildlife. Bisphenol A (BPA) is an estrogenic chemical commonly found in a variety of habitats. In this study, we used painted turtles (Chrysemys picta), which have temperature-dependent sex determination (TSD), as an animal model for ontogenetic endocrine disruption by BPA. We hypothesized that BPA would override TSD and disrupt sexual development. We incubated farm-raised turtle eggs at the male-producing temperature (26°C), randomly assigned individuals to treatment groups: control, vehicle control, 17β-estradiol (E2, 20ng/g-egg) or 0.01, 1.0, 100μgBPA/g-egg and harvested tissues at hatch. Typical female gonads were present in 89% of the E2-treated "males", but in none of the control males (n=35). Gonads of BPA-exposed turtles had varying amounts of ovarian-like cortical (OLC) tissue and disorganized testicular tubules in the medulla. Although the percentage of males with OLCs increased with BPA dose (BPA-low=30%, BPA-medium=33%, BPA-high=39%), this difference was not significant (p=0.85). In all three BPA treatments, SOX9 patterns revealed disorganized medullary testicular tubules and β-catenin expression in a thickened cortex. Liver vitellogenin, a female-specific liver protein commonly used as an exposure biomarker, was not induced by any of the treatments. Notably, these results suggest that developmental exposure to BPA disrupts sexual differentiation in painted turtles. Further examination is necessary to determine the underlying mechanisms of sex reversal in reptiles and how these translate to EDC exposure in wild populations.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC