Merck
OmpW is a potential target for eliciting protective immunity against Acinetobacter baumannii infections.
Vaccine (2015-07-25)
Weiwei Huang, Shijie Wang, Yufeng Yao, Ye Xia, Xu Yang, Qiong Long, Wenjia Sun, Cunbao Liu, Yang Li, Yanbing Ma
ABSTRAKT
Acinetobacter baumannii (A. baumannii) is an important conditioned pathogen that causes nosocomial and community-associated infections. In this study, we sought to investigate whether outer membrane protein W (OmpW) is a potential target for eliciting protective immunity against A. baumannii infections. Mice immunized with the fusion protein thioredoxin-OmpW generated strong OmpW-specific IgG responses. In a sepsis model, both active and passive immunizations against OmpW effectively protected mice from A. baumannii infections. This protection was demonstrated by a significantly improved survival rate, reduced bacterial burdens within organs, and the suppressed accumulation of inflammatory cytokines and chemokines in sera. Opsonophagocytic assays with murine macrophage RAW264.7 cells indicated that the bactericidal effects of the antisera derived from the immunized mice are mediated synergistically by specific antibodies and complement components. The antisera presented significant opsonophagocytic activities against homologous strains and clonally distinct clinical isolates in vitro. Protein data analysis showed that the sequence of OmpW, which has a molecule length of 183 amino acids, is more than 91% conserved in reported A. baumannii strains. In conclusion, we identified OmpW as a highly immunogenic and conserved protein as a valuable antigen candidate for the development of an effective vaccine or the preparation of antisera to control A. baumannii infections.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Orotidine 5′-monophosphate trisodium salt, ≥99% (HPLC), powder