Merck
  • A novel and green process for the production of tin oxide quantum dots and its application as a photocatalyst for the degradation of dyes from aqueous phase.
A novel and green process for the production of tin oxide quantum dots and its application as a photocatalyst for the degradation of dyes from aqueous phase.
Journal of colloid and interface science (2015-03-01)
Archita Bhattacharjee, M Ahmaruzzaman
ABSTRAKT
Green synthesis of SnO2 quantum dots (QDs) was developed by microwave heating method using the amino acids, namely, aspartic and glutamic acid. This method resulted in the formation of spherical SnO2 quantum dots with an average diameter less than the exciton Bohr radius of SnO2. The average diameter of SnO2 quantum dots formed using glutamic acid is ∼1.6 nm and is smaller than that formed using aspartic acid (∼2.6 nm). In the electronic spectra, a clear blue shift in the band gap energy from 4.33 to 4.4 eV is observed with a decrease in particle size (2.6-1.6 nm) due to three dimensional quantum confinement effects. The synthesized SnO2 QDs were characterized by transmission electron microscopy (TEM), selected area electron diffraction (SAED), X-ray diffraction (XRD) and Fourier transformed infrared spectroscopy (FT-IR). The optical properties were investigated using UV-visible spectroscopy. The synthesized SnO2 QDs act as an efficient photocatalyst in the degradation of Rose Bengal and Eosin Y dye under direct sunlight. For the first time, Rose Bengal dye was degraded using SnO2 QDs as a photocatalyst by solar irradiation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Eosin Y solution, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Sigma-Aldrich
Eosin Y disodium salt, ≥85 % by HPLC
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
Eosin Y disodium salt, certified by the BSC
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(II) chloride dihydrate, ≥99.97% trace metals basis