Merck
Bioaugmentation with isolated strains for the removal of toxic and refractory organics from coking wastewater in a membrane bioreactor.
Biodegradation (2015-10-30)
Xiaobiao Zhu, Rui Liu, Cong Liu, Lujun Chen
ABSTRAKT
The bioaugmentation strains for phenol, pyridine, quinoline, carbazole, and naphthalene degradation were employed to treat coking wastewater in a membrane bioreactor (MBR). The results showed that the bioaugmented MBR was much better in pollutant removal than that of the control MBR with conventional activated sludge. Compared to the control MBR, the bioaugmented MBR displayed an additional 3.2 mg/L of phenol, pyridine, quinoline, naphthalene and carbazole in total by the addition of the degrading strains. Also, about 10 % of the chemical oxygen demand in the effluent was further removed by the bioaugmentation. The pyrosequencing analysis of the sludge in the MBRs revealed that the microbial community shifted in response to the addition of the degrading strains. The diversity of the microbial community increased during the bioaugmentation, and some bacterial taxa favorable to the removal of toxic and refractory pollutants appeared in the bioaugmented MBR. The results indicated that the use of high-efficiency bacteria was a feasible method for industrial coking wastewater treatment.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
5,6-Dichlorobenzimidazole 1-β-D-ribofuranoside
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Carbazole, ≥95% (GC)
Sigma-Aldrich
Naphthalene, 99%
Sigma-Aldrich
Naphthalene, suitable for scintillation, ≥99%
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Quinoline, reagent grade, 98%
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)