Merck
  • Consequences of the loss of the Grainyhead-like 1 gene for renal gene expression, regulation of blood pressure and heart rate in a mouse model.
Consequences of the loss of the Grainyhead-like 1 gene for renal gene expression, regulation of blood pressure and heart rate in a mouse model.
Acta biochimica Polonica (2015-04-23)
Magdalena Pawlak, Agnieszka Walkowska, Michał Mlącki, Jelena Pistolic, Tomasz Wrzesiński, Vladimir Benes, Stephen M Jane, Joanna Wesoły, Elżbieta Kompanowska-Jezierska, Tomasz Wilanowski
ABSTRAKT
The Grainyhead-like 1 (GRHL1) transcription factor is tissue-specific and is very highly expressed in the kidney. In humans the GRHL1 gene is located at the chromosomal position 2p25. A locus conferring increased susceptibility to essential hypertension has been mapped to 2p25 in two independent studies, but the causative gene has never been identified. Furthermore, a statistically significant association has been found between a polymorphism in the GRHL1 gene and heart rate regulation. The aim of our study was to investigate the physiological consequences of Grhl1 loss in a mouse model and ascertain whether Grhl1 may be involved in the regulation of blood pressure and heart rate. In our research we employed the Grhl1 "knock-out" mouse strain. We analyzed renal gene expression, blood pressure and heart rate in the Grhl1-null mice in comparison with their "wild-type" littermate controls. Most important results: The expression of many genes is altered in the Grhl1(-/-) kidneys. Some of these genes have previously been linked to blood pressure regulation. Despite this, the Grhl1-null mice have normal blood pressure and interestingly, increased heart rate. Our work did not discover any new evidence to suggest any involvement of Grhl1 in blood pressure regulation. However, we determined that the loss of Grhl1 influences the regulation of heart rate in a mouse model.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Diethyl 1,4-dihydro-2,4,6-trimethyl-3,5-pyridinedicarboxylate, 99%
Sigma-Aldrich
Dibutyl phthalate, 99%
Sigma-Aldrich
DBP, 98% (HPLC)
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)