Merck
  • Synthesis of "click" alginate hydrogel capsules and comparison of their stability, water swelling, and diffusion properties with that of Ca(+2) crosslinked alginate capsules.
Synthesis of "click" alginate hydrogel capsules and comparison of their stability, water swelling, and diffusion properties with that of Ca(+2) crosslinked alginate capsules.
Journal of biomedical materials research. Part B, Applied biomaterials (2014-10-07)
Joyce C Breger, Benjamin Fisher, Raghu Samy, Steven Pollack, Nam Sun Wang, Irada Isayeva
ABSTRAKT
Ionically crosslinked alginate hydrogels have been extensively explored for encapsulation and immunoisolation of living cells/tissues to develop implantable cell therapies, such as islet encapsulation for bioartificial pancreas. Chemical instability of these hydrogels during long-term implantation hinders the development of viable cell therapy. The exchange between divalent crosslinking ions (e.g., Ca(+2) ) with monovalent ions from physiological environment causes alginate hydrogels to degrade, resulting in exposure of the donor tissue to the host's immune system and graft failure. The goal of this study was to improve stability of alginate hydrogels by utilizing covalent "click" crosslinking while preserving other biomedically viable hydrogel properties. Alginate was first functionalized to contain either pendant alkyne or azide functionalities, and subsequently reacted via "click" chemistry to form "click" gel capsules. Alginate functionalization was confirmed by NMR and gel permeation chromatography. When compared with Ca(+2) capsules, "click" capsules exhibited superior stability in ionic media, while showing higher permeability to small size diffusants and similar molecular weight cut-off and water swelling. Physicochemical properties of "click" alginate hydrogels demonstrate their potential utility for therapeutic cell encapsulation and other biomedical applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
11-Azido-3,6,9-trioxaundecan-1-amine, technical, ≥90% (GC)
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Propargylamine, 98%
Sigma-Aldrich
MES solution, BioUltra, Molecular Biology, 0.5 M in H2O
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
MES, low moisture content, ≥99% (titration)