Merck
Sc2O@Td 19151)-C76 : hindered cluster motion inside a tetrahedral carbon cage probed by crystallographic and computational studies.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-06-20)
Ting Yang, Yajuan Hao, Laura Abella, Qiangqiang Tang, Xiaohong Li, Yingbo Wan, Antonio Rodríguez-Fortea, Josep M Poblet, Lai Feng, Ning Chen
ABSTRAKT
A new cluster fullerene, Sc2 O@Td (19151)-C76 , has been isolated and characterized by mass spectrometry, UV/Vis/NIR absorption, (45) Sc NMR spectroscopy, cyclic voltammetry, and single-crystal X-ray diffraction. The crystallographic analysis unambiguously assigned the cage structure as Td (19151)-C76 , which is the first tetrahedral fullerene cage characterized by single-crystal X-ray diffraction. This study also demonstrated that the Sc2 O cluster has a much smaller ScOSc angle than that of Sc2 O@Cs (6)-C82 and the Sc2 O unit is fully ordered inside the Td (19151)-C76 cage. Computational studies further revealed that the cluster motion of the Sc2 O is more restrained in the Td (19151)-C76 cage than that in the Cs (6)-C82 cage. These results suggest that cage size affects not only the shapes but also the cluster motion inside fullerene cages.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Rhein
Sigma-Aldrich
Rhein, technical grade
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Benzene, anhydrous, 99.8%
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt