Merck
Electromembrane Extraction of Organic Acid Compounds in Biological Samples Followed by High-Performance Liquid Chromatography.
Journal of chromatographic science (2015-02-26)
Mostafa Khajeh, Mohammad Shakeri, Zahra Bameri Natavan, Zahra Safaei Moghaddam, Mousa Bohlooli, A A Moosavi-Movahedi
ABSTRAKT
Electromembrane extraction (EME) coupled with high-performance liquid chromatography was developed for determination of organic compounds including citric, tartaric and oxalic acid in biological samples. Organic compounds moved from aqueous samples, through a thin layer of 1-octanol immobilized in the pores of a porous hand-made polypropylene tube, and into a basic aqueous acceptor solution present inside the lumen of the tube. This new set-up for EME has a future potential such as simple, cheap and fast sample preparation technique for extraction of organic compounds in various complicated matrices. The pH of acceptor phase, extraction time, voltage, ionic strength, temperature and stirring speed were studied and optimized. Optimum conditions were: the pH of acceptor phase, 7; extraction time, 30 min; voltage, 30 V and stirring speed, 500 rpm. At the optimum conditions, the preconcentration factors of 175-200, the limits of detection of 1.9-3.1 µg L(-1) were obtained for the analytes. The developed procedure was then applied to the extraction and determination of organic acid compounds from biological samples.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Oxalic acid, puriss. p.a., anhydrous, ≥99.0% (RT)
Sigma-Aldrich
Oxalic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Oxalic acid, purified grade, 99.999% trace metals basis
Sigma-Aldrich
Oxalic acid, 98%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Citric acid, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)