Merck
Design, synthesis and antitubercular activity of certain nicotinic Acid hydrazides.
Molecules (Basel, Switzerland) (2015-05-20)
Wagdy M Eldehna, Mohamed Fares, Marwa M Abdel-Aziz, Hatem A Abdel-Aziz
ABSTRAKT
Three series of 6-aryl-2-methylnicotinohydrazides 4a-i, N'-arylidene-6-(4-bromophenyl)-2-methylnicotino hydrazides 7a-f, and N'-(un/substituted 2-oxoindolin-3-ylidene)-6-(4-fluorophenyl)-2-methylnicotinohydrazides 8a-c were synthesized and evaluated for their potential in vitro antimycobacterial activity against M. tuberculosis. The results showed that isatin hydrazides 8a-c are remarkably more active than the parent hydrazide 4c. Hydrazides 8b and 8c exhibited the highest activity among all the tested compounds (MIC = 12.5 and 6.25 µg/mL, respectively). Compounds 8b and 8c were also devoid of apparent cytotoxicity to HT-29, PC-3, A549, HepG2 and MCF-7 cancer cell lines. Besides, 8b and 8c showed good drug-likeness scores of 0.62 and 0.41, respectively. Those two isatin hydrazides could offer an excellent framework for future development to obtain more potent antitubercular agents. The SAR study suggested that lipophilicity of the synthesized derivatives is a crucial element that accounts for their antimycobacterial activity. Finally, a theoretical kinetic study was established to predict the ADME of the active derivatives.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Potassium bromide, powder, 99.95% trace metals basis
Sigma-Aldrich
Potassium bromide, powder, 99.999% trace metals basis