Merck
Nanodiscs as a Modular Platform for Multimodal MR-Optical Imaging.
Bioconjugate chemistry (2015-04-02)
Christiane E Carney, Ivan L Lenov, Catherine J Baker, Keith W MacRenaris, Amanda L Eckermann, Stephen G Sligar, Thomas J Meade
ABSTRAKT
Nanodiscs are monodisperse, self-assembled discoidal particles that consist of a lipid bilayer encircled by membrane scaffold proteins (MSP). Nanodiscs have been used to solubilize membrane proteins for structural and functional studies and deliver therapeutic phospholipids. Herein, we report on tetramethylrhodamine (TMR) tagged nanodiscs that solubilize lipophilic MR contrast agents for generation of multimodal nanoparticles for cellular imaging. We incorporate both multimeric and monomeric Gd(III)-based contrast agents into nanodiscs and show that particles containing the monomeric agent (ND2) label cells with high efficiency and generate significant image contrast at 7 T compared to nanodiscs containing the multimeric agent (ND1) and Prohance, a clinically approved contrast agent.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycero-3-phosphocholine, ≥99%
Sigma-Aldrich
3,3′-Diiodo-L-thyronine (T2) hydrochloride, 98% (CP)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture