Merck
  • Microbial invasion and histological chorioamnionitis upregulate neutrophil-gelatinase associated lipocalin in preterm prelabor rupture of membranes.
Microbial invasion and histological chorioamnionitis upregulate neutrophil-gelatinase associated lipocalin in preterm prelabor rupture of membranes.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians (2014-11-27)
Marie Vajrychova, Marian Kacerovsky, Vojtech Tambor, Helena Hornychova, Juraj Lenco
ABSTRAKT
Our recent exploratory proteomic study suggested increased levels of neutrophil-gelatinase associated lipocalin (P80188, NGAL_HUMAN) due to microbial invasion of the amniotic cavity (MIAC) and histological chorioamnionitis (HCA) in women with preterm prelabor rupture of the membranes. In this study, we verified the proteomics findings by assessing the amniotic fluid NGAL by ELISA in the original exploratory cohort. The NGAL level was significantly higher in women positive for both MIAC and HCA compared to women with both conditions ruled out (median 75.1 ng/ml versus 27.9 ng/ml; p < 0.0001). For independent validation and to assess NGALs potential to stratify women positive for both MIAC and HCA from women in whom at least one of these conditions was absent, we subsequently designed a retrospective replication cohort. Significantly higher NGAL levels were found in women positive for both MIAC and HCA (median 65.9 ng/ml versus 34.2 ng/ml; p = 0.0061). Significantly higher levels of NGAL were confirmed only in strata below 32 weeks of gestation. Based on the observed likelihood ratio, the best predictive cutoff level (47.1 ng/ml) was evaluated in both cohorts. Data from the verification cohort implied that NGAL is a valuable clinical marker for revealing MIAC leading to HCA; however, this potential was not replicated in the replication cohort.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder