Merck
GC-MS method for determining faecal sterols as biomarkers of human and pastoral animal presence in freshwater sediments.
Analytical and bioanalytical chemistry (2015-09-08)
Dario Battistel, Rossano Piazza, Elena Argiriadis, Enrico Marchiori, Marta Radaelli, Carlo Barbante
ABSTRAKT
In order to determine sterols and stanols in freshwater sediments to reconstruct the past presence of humans and pastoral animals, we developed an analytical method based on pressurised liquid extraction (PLE), clean-up performed using solid phase extraction (SPE) and sterol determination using gas chromatography-mass spectrometry (GC-MS) analysis. PLE extraction conditions were optimised using dichloromethane (DCM) and DCM/methanol mixtures. Clean-up was performed with 2 g silica SPE cartridges, and the concentrated extracts were eluted with 70 mL DCM. Extraction yield was evaluated using an in-house reference material spiked with (13)C-labelled cholesterol and aged for 10 days. In comparison with pre-extraction, where the sediment is extracted and then spiked with a known analyte concentration, this approach preserves the original composition of the sediment. DCM and DCM/methanol mixtures resulted in high extraction yields ranging from 86 to 92 % with good reproducibility (relative standard deviation (RSD) 5-8 %). PLE extraction yields obtained with DCM as the extracting solvent were about 1.5 times higher than extractions using an ultrasonic bath. The solvent extraction mixture and matrix composition strongly affected the solvent extraction composition where higher overall recoveries (70-80 %) for each compound were obtained with DCM. The extraction mixture and matrix composition also affected the analyte concentrations, resulting in a method precision ranging from 1 to 18 %. Diatomaceous earth spiked with 10 to 100 ng of sterols, and environmental samples fortified with suitable amounts of sterols provided apparent recovery values ranging from 90 to 110 %. We applied the method to environmental samples both close to and upstream from sewage discharge zones, resulting in substantially higher faecal sterol (FeSt) concentrations near the sewage. In addition, we also applied the method to a 37-cm freshwater sediment core in order to evaluate its applicability for obtaining vertical sterol profiles.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Sodium sulfate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Sodium sulfate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, powder
Sigma-Aldrich
Sodium sulfate, ACS reagent, ≥99.0%, anhydrous, granular
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer