Merck
High selective antileishmanial activity of vanadium complex with stilbene derivative.
Acta tropica (2015-04-29)
Patrícia de Almeida Machado, Vinícius Zamprogno Mota, Ana Clara de Lima Cavalli, Gustavo Senra Gonçalves de Carvalho, Adilson David Da Silva, Jacy Gameiro, Alexandre Cuin, Elaine Soares Coimbra
ABSTRAKT
Leishmaniasis is a group of disease caused by different species of the parasite Leishmania affecting millions of people worldwide. Conventional therapy relies on multiple parenteral injections with pentavalent antimonials which exhibit high toxicity and various side effects have been reported. Hence, the research for an effective and low toxic effect drug is necessary. In the present work, the synthesis, spectroscopic and analytical characterizations of stilbene derivative (H2Salophen) and its vanadium complex (VOSalophen) are reported. Besides the chemical ancillary information, investigation of the leishmanicidal effects of these compounds were provided. The biological assays against promastigote and amastigote forms of L. amazonensis have been shown that VOSalophen exhibited a strong antiparasitic activity (IC50 of 6.65 and 3.51 μM, respectively). Furthermore, the leishmanicidal activity was concentration and time-dependent. Regarding toxicity and selectivity on mammalian cells, VOSalophen have not caused significant damage to human erythrocytes in all concentrations tested and VOSalophen was almost seven times more destructive for the intracellular parasite than for macrophages. Furthermore, the leishmanicidal activity of VOSalophen in promastigote forms of L. amazonensis could be associated to mitochondrial dysfunction and increase of the reactive oxygen species (ROS) production. In L. amazonensis-infected macrophages, VOSalophen induces ROS production and a microbicidal action NO-dependent. Our biological results indicate the effective and selective action of VOSalophen against L. amazonensis and the leishmanicidal effect can be associated to parasite disorders and immumodulatory effects.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ≥98%
Sigma-Aldrich
Vanadium, powder, -100 mesh, 99.9% trace metals basis
Sigma-Aldrich
o-Phenylenediamine, sublimed, ≥99%
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Dioctan 2′,7′-dichlorodihydrofluoresceiny, ≥97%
Sigma-Aldrich
o-Phenylenediamine, Peroxidase substrate, ≥98.0%, powder
Sigma-Aldrich
o-Phenylenediamine, tablet, 20 mg substrate per tablet
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Sulfanilamide, ≥98%
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Sigma-Aldrich
Vanadium, pieces, 1-3 mm, 99.9% trace metals basis
Sigma-Aldrich
Salicylaldehyde, redist., ≥99.0% (GC)
Sigma-Aldrich
Salicylaldehyde, ≥98%, FG
Sigma-Aldrich
Salicylaldehyde, reagent grade, 98%
Sigma-Aldrich
Sulfanilamide, puriss. p.a., ≥98% (calc. to the dried substance)
Vanadium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.8%
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Phosphoric acid solution, suitable for NMR (reference standard), 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
Sigma-Aldrich
o-Phenylenediamine, flaked, 99.5%
Sigma-Aldrich
N-(1-Naphthyl)ethylenediamine dihydrochloride, ACS reagent, >98%
Sigma-Aldrich
Miltefosine, ≥98% (perchloric acid titration)
Sigma-Aldrich
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
Vanadium, wire reel, 0.2m, diameter 0.5mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 0.2m, diameter 1.0mm, Stress relieved, 99.8%
Vanadium, wire reel, 1m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.2m, diameter 0.1mm, as drawn, 99.8%
Vanadium, wire reel, 0.5m, diameter 0.25mm, hard, 99.8%