Merck
Is microdialysis useful for early detection of acute rejection after kidney transplantation?
International journal of surgery (London, England) (2015-04-14)
Hamidreza Fonouni, Mohammad Golriz, Ali Majlesara, Alireza Faridar, Majid Esmaeilzadeh, Parvin Jarahian, Morva Tahmasbi Rad, Mohammadreza Hafezi, Camelia Garoussi, Stephan Macher-Goeppinger, Thomas Longerich, Berk Orakcioglu, Oliver W Sakowitz, Arianeb Mehrabi
ABSTRAKT
Acute rejection following kidney transplantation (KTx) is still one of the challenging complications leading to chronic allograft failure. The aim of this study was to investigate the role of microdialysis (MD) in the early detection of acute graft rejection factor following KTx in porcine model. Sixteen pigs were randomized after KTx into case (n = 8, without immunosuppressant) and control groups (n = 8, with immunosuppressant). The rejection diagnosis in our groups was confirmed by histopathological evidences as "acute borderline rejection". Using MD, we monitored the interstitial concentrations of glucose, lactate, pyruvate, glutamate and glycerol in the transplanted grafts after reperfusion. In the early post-reperfusion phase the lactate level in our case group was significantly higher comparing to the control group and remained in higher levels until the end of monitoring. The lactate to pyruvate ratio showed a considerable increase in the case group during the post-reperfusion phase. The other metabolites (glucose, glycerol, glutamate) were nearly at the same levels at the end of our monitoring in both study groups. The increase in lactate and lactate to pyruvate ratios seems to be an indicator for early detection of acute rejection after KTx. Therefore, MD as a minimally invasive measurement tool may help to identify the need to immunosuppression adjustment in the early KTx phase before the clinical manifestation of the rejection.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
6α-Methylprednisolone, ≥98%