Merck
Phosphine-free synthesis of Ag-In-Se alloy nanocrystals with visible emissions.
Nanoscale (2015-10-23)
Dong Yao, Huiwen Liu, Yi Liu, Chunwei Dong, Kai Zhang, Yu Sheng, Jianli Cui, Hao Zhang, Bai Yang
ABSTRAKT
As promising heavy metal-free emitting materials, Ag-In-Se nanocrystals (NCs) are conventionally synthesized using organic phosphine agents and exhibit near-infrared emissions. In this work, we demonstrate a rapid phosphine-free approach for synthesizing Ag-In-Se alloy NCs with the emissions tunable to the visible region on the basis of the phosphine-free dissolution of Se powder. At room temperature, Se powder is reduced by dodecanethiol and dissolved in oleylamine to produce a Se precursor. The resultant Se precursor is highly active, which permits rapid synthesis at a relatively low temperature, such as at 90 °C for 150 s. By optimizing the size, structure, and composition, the photoluminescence quantum yield of the as-synthesized Ag-In-Se NCs is enhanced to up to 10%. The growth of the Ag-In-Se NCs involves composition and phase transition, which strongly depend on the reaction temperature. The Ag2Se nuclei form first, and the Ag-In-Se NCs are produced by doping In(3+) into the preformed Ag2Se nuclei. Tetragonal phase Ag-In-Se is obtained below 170 °C, while the orthorhombic phase appears over 190 °C. The potential of Ag-In-Se NCs as red emitting phosphors for lighting-emitting diodes is further demonstrated.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
1-Decanethiol, 99%
Sigma-Aldrich
1-Decanethiol, 96%
SAFC
Oleylamine, ≥98% (primary amine)
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
1-Dodecanethiol, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Oleylamine, technical grade, 70%