- Studies on the biosynthesis of ralfuranones in Ralstonia solanacearum.
Studies on the biosynthesis of ralfuranones in Ralstonia solanacearum.
Bioscience, biotechnology, and biochemistry (2015-12-10)
Kenji Kai, Hideyuki Ohnishi, Akinori Kiba, Kouhei Ohnishi, Yasufumi Hikichi
PMID26645956
ABSTRAKT
Ralfuranones, aryl-furanone secondary metabolites, are involved in the virulence of Ralstonia solanacearum in solanaceous plants. Ralfuranone I (6) has been suggested as a biosynthetic precursor for other ralfuranones; however, this conversion has not been confirmed. We herein investigate the biosynthesis of ralfuranones using feeding experiments with ralfuranone I (6) and its putative metabolite, ralfuranone B (2). The results obtained demonstrated that the biosynthesis of ralfuranones proceeded in enzymatic and non-enzymatic manners.
MATERIAŁY