Merck
  • β-Elemene induces apoptosis of human rheumatoid arthritis fibroblast-like synoviocytes via reactive oxygen species-dependent activation of p38 mitogen-activated protein kinase.
β-Elemene induces apoptosis of human rheumatoid arthritis fibroblast-like synoviocytes via reactive oxygen species-dependent activation of p38 mitogen-activated protein kinase.
Pharmacological reports : PR (2016-01-02)
Shibo Zou, Changyin Wang, Zhengjun Cui, Pengfei Guo, Qingnan Meng, Xun Shi, Ya Gao, Gaoyuan Yang, Zhaofeng Han
ABSTRAKT
β-Elemene is a natural anticancer compound extracted from the Chinese medicinal herb Curcuma Wenyujin. This study was done to determine the effect of β-elemene on the apoptosis of rheumatoid arthritis fibroblast-like synoviocytes (RA-FLS) and associated molecular mechanisms. RA-FLS were treated for 72h with β-elemene at 10-200μg/ml and cell viability and apoptotic changes were examined. The involvement of reactive oxygen species (ROS) and mitogen-activated protein kinases (MAPKs) was checked. We found that β-elemene significantly inhibited the viability and promoted apoptosis of RA-FLS in a concentration-dependent fashion. β-Elemene-treated FLS showed a significant decline in mitochondrial membrane potential, an accumulation of cytochrome c in the cytosol, and increased activities of caspase-9 and caspase-3. β-Elemene treatment caused an enhancement of p38 MAPK phosphorylation and ROS production. The pro-apoptotic activity of β-elemene was significantly reversed by pretreatment with the p38 inhibitor SB203580 or ROS inhibitor N-acetyl-l-cysteine. Taken together, β-elemene is effective in inducing mitochondrial apoptosis of RA-FLS, which is mediated through induction of ROS formation and p38 MAPK activation. β-Elemene may thus have therapeutic benefits for RA.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DCFHDA, BioReagent, suitable for fluorescence, ≥95% (HPLC)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)