Merck
Oxidative stress-induced apoptosis of endothelial cells.
Free radical biology & medicine (2000-10-12)
M C Warren, E A Bump, D Medeiros, S J Braunhut
ABSTRAKT
Endothelial cells (ECs) are subjected to oxidative stress during many pathological processes, including ischemia/reperfusion and general inflammation. In the present study, we examined the effects of oxidative stress on rates of apoptosis in EC cultures. We treated large and microvessel ECs with menadione for 1 h in vitro to simulate the most common physiological form of oxidative stress, exposure to O2*-. Capillary ECs were resistant to menadione-induced apoptosis when compared with large-vessel ECs. Treatment with 35 microM menadione resulted in an apoptotic rate of approximately 5% in capillary EC cultures compared with approximately 45% in large-vessel EC cultures. At higher concentrations of menadione (35-75 microM), both types of ECs exhibited a concentration-related increase in apoptosis. Necrotic cell death only became evident at menadione concentrations ranging from 75-100 microM for both cell types. The timing of the apoptotic response to a 1 h menadione exposure was very specific. For both EC types, peaks of apoptosis occurred in two distinct waves, at 6-8 and 18-22 h after treatment. Analysis of the events leading up to the first peak of apoptosis indicated that specific matrix metalloproteinases (MMPs) were activated, suggesting that MMPs may be involved in initiating the apoptotic process.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Menadione, crystalline