Merck
The 5'-AMP-Activated Protein Kinase (AMPK) Is Involved in the Augmentation of Antioxidant Defenses in Cryopreserved Chicken Sperm.
PloS one (2015-07-30)
Thi Mong Diep Nguyen, François Seigneurin, Pascal Froment, Yves Combarnous, Elisabeth Blesbois
ABSTRAKT
Semen cryopreservation is a unique tool for the management of animal genetic diversity. However, the freeze-thaw process causes biochemical and physical alterations which make difficult the restoration of sperm energy-dependent functions needed for fertilization. 5'-AMP activated protein kinase (AMPK) is a key sensor and regulator of intracellular energy metabolism. Mitochondria functions are known to be severely affected during sperm cryopreservation with deleterious oxidative and peroxidative effects leading to cell integrity and functions damages. The aim of this study was thus to examine the role of AMPK on the peroxidation/antioxidant enzymes defense system in frozen-thawed sperm and its consequences on sperm functions. Chicken semen was diluted in media supplemented with or without AMPK activators (AICAR or Metformin [MET]) or inhibitor (Compound C [CC]) and then cryopreserved. AMPKα phosphorylation, antioxidant enzymes activities, mitochondrial potential, ATP, citrate, viability, acrosome reaction ability (AR) and various motility parameters were negatively affected by the freeze-thaw process while reactive oxygen species (ROS) production, lipid peroxidation (LPO) and lactate concentration were dramatically increased. AICAR partially restored superoxide dismutase (SOD), Glutathione Peroxidase (GPx) and Glutathione Reductase (GR), increased ATP, citrate, and lactate concentration and subsequently decreased the ROS and LPO (malondialdehyde) in frozen-thawed semen. Motility parameters were increased (i.e., + 23% for motility, + 34% for rapid sperm) as well as AR (+ 100%). MET had similar effects as AICAR except that catalase activity was restored and that ATP and mitochondrial potential were further decreased. CC showed effects opposite to AICAR on SOD, ROS, LPO and AR and motility parameters. Taken together, our results strongly suggest that, upon freeze-thaw process, AMPK stimulated intracellular anti-oxidative defense enzymes through ATP regulation, thus reducing ROS and lipid peroxidation, and consequently partially restoring several essential sperm functions and leading to a better quality of cryopreserved sperm.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder