Merck
Role of microRNAs Located on Chromosome Arm 10q in Malignant Gliomas.
Brain pathology (Zurich, Switzerland) (2015-08-01)
Marietta Wolter, Thomas Werner, Bastian Malzkorn, Guido Reifenberger
ABSTRAKT
Deletions of chromosome arm 10q are found in most glioblastomas and subsets of lower grade gliomas. Mutations in the PTEN gene at 10q23.3 are restricted to less than half of the 10q-deleted gliomas, suggesting additional glioma-associated tumor suppressors on 10q. We investigated 64 astrocytic gliomas of different malignancy grades for aberrant expression of 16 microRNAs (miRNAs) on 10q. Thereby, we identified four miRNAs (miR-107, miR-146b-5p, miR-346, miR-1287-5p) whose expression was frequently down-regulated in anaplastic astrocytomas and/or glioblastomas. DNA methylation analyses revealed 5'-CpG site hypermethylation of miR-346 in more than two-thirds of primary glioblastomas, while aberrant 5'-CpG site methylation of miR-146b-5p was frequent in IDH1-mutant astrocytomas and secondary glioblastomas. Overexpression of either of the four miRNAs in glioma cell lines reduced cell proliferation and/or increased caspase-3/7 activity. Expression analyses of miRNA overexpressing glioma cells and 3'-untranslated region luciferase reporter gene assays revealed evidence that these miRNAs post-transcriptionally regulate expression of glioma-relevant genes, including CDK6 (miR-107), EGFR (miR-146b-5p, miR-1287-5p), TERT and SEMA6A (miR-346), all of which are overexpressed in malignant gliomas in situ. In summary, we show that the 10q-located miRNAs miR-107, miR-146b-5p, miR-346 and miR-1287-5p are frequently down-regulated in malignant gliomas and thereby may support overexpression of important glioma growth-promoting genes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid