Merck
Activation of Nicotinic Acetylcholine Receptors Decreases Apoptosis in Human and Female Murine Pancreatic Islets.
Endocrinology (2016-07-30)
Philippe Klee, Domenico Bosco, Audrey Guérardel, Emmanuel Somm, Audrey Toulotte, Pierre Maechler, Valérie M Schwitzgebel
ABSTRAKT
Type 1 diabetes (T1DM) results from destruction of most insulin-secreting pancreatic β-cells. The persistence of β-cells decades after the onset of the disease indicates that the resistance of individual cells to the autoimmune insult is heterogeneous and might depend on the metabolic status of a cell at a given moment. The aim of this study is to investigate whether activation of nicotinic acetylcholine receptors (nACh-Rs) could increase β-cell resistance against the adverse environment prevailing at the onset of T1DM. Here, we show that nACh-R activation by nicotine and choline, 2 agonists of the receptor, decreases murine and human β-cell apoptosis induced by proinflammatory cytokines known to be present in the islet environment at the onset of T1DM. The protective mechanism activated by nicotine and choline involves attenuation of mitochondrial outer membrane permeabilization via modulation of endoplasmic reticulum stress, of the activity of B-cell lymphoma 2 family proteins and cytoplasmic calcium levels. Local inflammation and endoplasmic reticulum stress being key determinants of β-cell death in T1DM, we conclude that pharmacological activation of nACh-R could represent a valuable therapeutic option in the modulation of β-cell death in T1DM.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-Insulin antibody produced in mouse, clone K36AC10, ascites fluid
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%