Merck
Polyphosphates and Fulvates Enhance Environmental Stability of PO
Journal of agricultural and food chemistry (2016-10-19)
Jessica Bollyn, Mathias Nijsen, Stijn Baken, Iris Joye, Nadia Waegeneers, Geert Cornelis, Erik Smolders
ABSTRAKT
Iron oxyhydroxide nanoparticles (Fe-NPs) are natural vectors of phosphate (PO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
meso-Tetraphenylporphyrin, BioReagent, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Trisodium trimetaphosphate, ≥95%