Merck
CRIS-a novel cAMP-binding protein controlling spermiogenesis and the development of flagellar bending.
PLoS genetics (2013-12-18)
Anke Miriam Krähling, Luis Alvarez, Katharina Debowski, Qui Van, Monika Gunkel, Stephan Irsen, Ashraf Al-Amoudi, Timo Strünker, Elisabeth Kremmer, Eberhard Krause, Ingo Voigt, Simone Wörtge, Ari Waisman, Ingo Weyand, Reinhard Seifert, Ulrich Benjamin Kaupp, Dagmar Wachten
ABSTRAKT
The second messengers cAMP and cGMP activate their target proteins by binding to a conserved cyclic nucleotide-binding domain (CNBD). Here, we identify and characterize an entirely novel CNBD-containing protein called CRIS (cyclic nucleotide receptor involved in sperm function) that is unrelated to any of the other members of this protein family. CRIS is exclusively expressed in sperm precursor cells. Cris-deficient male mice are either infertile due to a lack of sperm resulting from spermatogenic arrest, or subfertile due to impaired sperm motility. The motility defect is caused by altered Ca(2+) regulation of flagellar beat asymmetry, leading to a beating pattern that is reminiscent of sperm hyperactivation. Our results suggest that CRIS interacts during spermiogenesis with Ca(2+)-regulated proteins that--in mature sperm--are involved in flagellar bending.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało przeciw fosfotyrozynie, klon ®, clone 4G10®, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Tubulin antibody produced in mouse, clone TUB 2.1, ascites fluid
Sigma-Aldrich
Anti-Calnexin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution