Merck
Attenuation of lithium-induced natriuresis and kaliuresis in P2Y₂ receptor knockout mice.
American journal of physiology. Renal physiology (2013-06-07)
Yue Zhang, Lijun Li, Donald E Kohan, Carolyn M Ecelbarger, Bellamkonda K Kishore
ABSTRAKT
Whole body knockout (KO) of the P2Y₂ receptor (P2Y₂R) results in enhanced vasopressin V2 receptor activity and increased renal Na⁺ conservation. We hypothesized that P2Y₂R KO mice would be less sensitive to lithium-induced natriuresis and kaliuresis due to attenuated downregulation of one or more of the major renal Na⁺ or K⁺ transporter/channel proteins. KO and wild-type (WT) mice were fed a control or lithium-added diet (40 mmol/kg food) for 14 days. Lithium-induced natriuresis and kaliuresis were significantly (~25%) attenuated in KO mice. The subunits of the epithelial Na⁺ channel (ENaC) were variably affected by lithium and genotype, but, overall, medullary levels were decreased substantially by lithium (15-60%) in both genotypes. In contrast, cortical, β-, and γ-ENaC were increased by lithium (~50%), but only in WT mice. Moreover, an assessment of ENaC activity by benzamil sensitivity suggested that lithium increased ENaC activity in WT mice but in not KO mice. In contrast, medullary levels of Na⁺-K⁺-2Cl⁻ cotransporter 2 and cortical levels of the renal outer medullary K⁺ channel were not downregulated by lithium and were significantly (15-76%) higher in KO mice under both dietary conditions. In addition, under control conditions, tissue osmolality of the inner medulla as well as furosemide sensitivity were significantly higher in KO mice versus WT mice. Therefore, we suggest that increased expression of these proteins, particularly in the control state, reduces Na⁺ delivery to the distal nephron and provides a buffer to attenuate collecting duct-mediated natriuresis and kaliuresis. Additional studies are warranted to explore the potential therapeutic benefits of purinergic antagonism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Przeciwciało anty-Na+/K+ ATPaza α-1, klon C464.6, clone C464.6, Upstate®, from mouse
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture