Merck
Involvement of an NKG2D ligand H60c in epidermal dendritic T cell-mediated wound repair.
Journal of immunology (Baltimore, Md. : 1950) (2012-03-10)
Shigeru Yoshida, Rania Hassan Mohamed, Mizuho Kajikawa, Jun Koizumi, Minami Tanaka, Kazunori Fugo, Noriyuki Otsuka, Katsumi Maenaka, Hideo Yagita, Hitoshi Chiba, Masanori Kasahara
ABSTRAKT
Dendritic epidermal T cells (DETCs) found in mouse skin are NKG2D-positive γδ T cells involved in immune surveillance and wound repair. It is assumed that the interaction of an NKG2D receptor on DETCs and an MHC class I-like NKG2D ligand on keratinocytes activates DETCs, which then secrete cytokines promoting wound repair. However, direct evidence that DETC activation through NKG2D signaling promotes wound repair is not available. In the present study, we generated mAbs for an NKG2D ligand H60c previously suggested to be expressed specifically on skin keratinocytes. Local administration of H60c-specific mAb inhibited activation of DETCs and significantly delayed wound repair. Likewise, administration of NKG2D-specific mAb impaired wound repair to a similar extent. The delay in wound closure resulting from the blockade of the NKG2D pathway was comparable to that observed in γδ T cell-deficient mice. These results indicate that H60c/NKG2D interactions play a critical role in wound repair. Reassessment of binding affinities showed that H60c monomers bind to NKG2D with affinity (K(d) = 26 ± 3.2 nM) comparable to those of other high-affinity NKG2D ligands. H60c is transcribed not only in skin but also in tissues such as tongue and female reproductive tract known to contain epithelium-resident γδ T cells expressing invariant TCRs, suggesting a more general role for H60c in the maintenance of epithelial integrity.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
IgG from rat serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution
Sigma-Aldrich
IgG from rabbit serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Millipore
ANTI-FLAG® antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution