Merck
Pim2 is required for maintaining multiple myeloma cell growth through modulating TSC2 phosphorylation.
Blood (2013-07-03)
Jing Lu, Tatiana Zavorotinskaya, Yumin Dai, Xiao-Hong Niu, Joseph Castillo, Janet Sim, Jianjun Yu, Yingyun Wang, John L Langowski, Jocelyn Holash, Kevin Shannon, Pablo D Garcia
ABSTRAKT
Multiple myeloma (MM) is the second most common hematologic malignancy. Despite recent treatment advances, it remains incurable. Here, we report that Pim2 kinase expression is highly elevated in MM cells and demonstrate that it is required for MM cell proliferation. Functional interference of Pim2 activity either by short hairpin RNAs or by a potent and selective small-molecule inhibitor leads to significant inhibition of MM cell proliferation. Pim inhibition results in a significant decrease of mammalian target of rapamycin C1 (mTOR-C1) activity, which is critical for cell proliferation. We identify TSC2, a negative regulator of mTOR-C1, as a novel Pim2 substrate and show that Pim2 directly phosphorylates TSC2 on Ser-1798 and relieves the suppression of TSC2 on mTOR-C1. These findings support Pim2 as a promising therapeutic target for MM and define a novel Pim2-TSC2-mTOR-C1 pathway that drives MM proliferation.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Przeciwciało anty-Raptor, klon 1H6.2, clone 1H6.2, from mouse
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder