Merck
Bruton's tyrosine kinase revealed as a negative regulator of Wnt-beta-catenin signaling.
Science signaling (2009-05-28)
Richard G James, Travis L Biechele, William H Conrad, Nathan D Camp, Daniel M Fass, Michael B Major, Karen Sommer, XianHua Yi, Brian S Roberts, Michele A Cleary, William T Arthur, Michael MacCoss, David J Rawlings, Stephen J Haggarty, Randall T Moon
ABSTRAKT
Wnts are secreted ligands that activate several receptor-mediated signal transduction cascades. Homeostatic Wnt signaling through beta-catenin is required in adults, because either elevation or attenuation of beta-catenin function has been linked to diverse diseases. To contribute to the identification of both protein and pharmacological regulators of this pathway, we describe a combinatorial screen that merged data from a high-throughput screen of known bioactive compounds with an independent focused small interfering RNA screen. Each screen independently revealed Bruton's tyrosine kinase (BTK) as an inhibitor of Wnt-beta-catenin signaling. Loss of BTK function in human colorectal cancer cells, human B cells, zebrafish embryos, and cells derived from X-linked agammaglobulinemia patients with a mutant BTK gene resulted in elevated Wnt-beta-catenin signaling, confirming that BTK acts as a negative regulator of this pathway. From affinity purification-mass spectrometry and biochemical binding studies, we found that BTK directly interacts with a nuclear component of Wnt-beta-catenin signaling, CDC73. Further, we show that BTK increased the abundance of CDC73 in the absence of stimulation and that CDC73 acted as a repressor of beta-catenin-mediated transcription in human colorectal cancer cells and B cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Monoclonal Anti-β-Tubulin I antibody produced in mouse, clone SAP.4G5, ascites fluid
Sigma-Aldrich
Monoclonal ANTI-FLAG® antibody produced in rabbit, clone SIG1-25, ascites fluid
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Roche
cOmplete, Mini, Koktajl inhibitorów proteaz bez EDTA, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial