Merck
Apoptosis is required during early stages of tail regeneration in Xenopus laevis.
Developmental biology (2006-12-08)
Ai-Sun Tseng, Dany S Adams, Dayong Qiu, Punita Koustubhan, Michael Levin
ABSTRAKT
The Xenopus tadpole is able to regenerate its tail, including skin, muscle, notochord, spinal cord and neurons and blood vessels. This process requires rapid tissue growth and morphogenesis. Here we show that a focus of apoptotic cells appears in the regeneration bud within 12 h of amputation. Surprisingly, when caspase-3 activity is specifically inhibited, regeneration is abolished. This is true of tails both before and after the refractory period. Programmed cell death is only required during the first 24 h after amputation, as later inhibition has no effect on regeneration. Inhibition of caspase-dependent apoptosis results in a failure to induce proliferation in the growth zone, a mispatterning of axons in the regenerate, and the appearance of ectopic otoliths in the neural tube, in the context of otherwise normal continued development of the larva. Larvae amputated during the refractory stage exhibit a much broader domain of caspase-3-positive cells, suggesting a window for the amount of apoptosis that is compatible with normal regeneration. These data reveal novel roles for apoptosis in development and indicate that a degree of apoptosis is an early and obligate component of normal tail regeneration, suggesting the possibility of the existence of endogenous inhibitory cells that must be destroyed by programmed cell death for regeneration to occur.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Inhibitor apoptozy II, NS3694, The Apoptosis Inhibitor II, NS3694, also referenced under CAS 426834-38-0, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Przeciwciało anty-fosfo-histonowe H3 (Ser10), klon RR002, clone RR002, Upstate®, from mouse
Sigma-Aldrich
Inhibitor apoptozy, The Apoptosis Inhibitor, also referenced under CAS 54135-60-3, controls the biological activity of Apoptosis. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid