Merck
Endothelium-dependent vasodilation effects of
Experimental and therapeutic medicine (2017-01-20)
Yanyan Wang, Yu Ren, Leilei Xing, Xiangdong Dai, Sheng Liu, Bin Yu, Yi Wang
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
1H-[1,2,4]Oxadiazolo[4,3-a]quinoxalin-1-one, powder
Sigma-Aldrich
Acetylcholine bromide, ≥99%