Merck
  • Intact wound repair activity of human mesenchymal stem cells after YM155 mediated selective ablation of undifferentiated human embryonic stem cells.
Intact wound repair activity of human mesenchymal stem cells after YM155 mediated selective ablation of undifferentiated human embryonic stem cells.
Journal of dermatological science (2017-02-12)
Keun-Tae Kim, Ho-Chang Jeong, C-Yoon Kim, Eun-Young Kim, Si-Hyun Heo, Seung-Ju Cho, Ki-Sung Hong, Hyuk-Jin Cha
ABSTRAKT
Risk of teratoma formation during human pluripotent stem cell (hPSC)-based cell therapy is one of the technical hurdles that must be resolved before their wider clinical application. To this end, selective ablation of undifferentiated hPSCs has been achieved using small molecules whose application should be safe for differentiated cells derived from the hPSCs. However, the functional safety of such small molecules in the cells differentiated from hPSCs has not yet been extensively validated. We used the survivin inhibitor YM155, which induced highly selective cell death of hPSCs for ablating undifferentiated hESCs after differentiation to human mesenchymal stem cells (hMSCs) and examined whether hMSCs remained fully functional after being exposed by YM155. We demonstrated that human mesenchymal stem cells (hMSCs) derived from human embryonic stem cells (hESCs) remained fully functional in vitro and in vivo, while hESCs were selectively ablated. These results suggest that a single treatment with YM155 after differentiation of hMSCs would be a valid approach for teratoma-free cell therapy.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Alizarin, Dye content 97 %