Merck
Induction of an IFN-Mediated Antiviral Response by a Self-Amplifying RNA Vaccine: Implications for Vaccine Design.
Journal of immunology (Baltimore, Md. : 1950) (2017-04-19)
Timothy Pepini, Anne-Marie Pulichino, Thomas Carsillo, Alicia L Carlson, Farid Sari-Sarraf, Katrin Ramsauer, Jason C Debasitis, Giulietta Maruggi, Gillis R Otten, Andrew J Geall, Dong Yu, Jeffrey B Ulmer, Carlo Iavarone
ABSTRAKT
RNA-based vaccines have recently emerged as a promising alternative to the use of DNA-based and viral vector vaccines, in part because of the potential to simplify how vaccines are made and facilitate a rapid response to newly emerging infections. SAM vaccines are based on engineered self-amplifying mRNA (SAM) replicons encoding an Ag, and formulated with a synthetic delivery system, and they induce broad-based immune responses in preclinical animal models. In our study, in vivo imaging shows that after the immunization, SAM Ag expression has an initial gradual increase. Gene expression profiling in injection-site tissues from mice immunized with SAM-based vaccine revealed an early and robust induction of type I IFN and IFN-stimulated responses at the site of injection, concurrent with the preliminary reduced SAM Ag expression. This SAM vaccine-induced type I IFN response has the potential to provide an adjuvant effect on vaccine potency, or, conversely, it might establish a temporary state that limits the initial SAM-encoded Ag expression. To determine the role of the early type I IFN response, SAM vaccines were evaluated in IFN receptor knockout mice. Our data indicate that minimizing the early type I IFN responses may be a useful strategy to increase primary SAM expression and the resulting vaccine potency. RNA sequence modification, delivery optimization, or concurrent use of appropriate compounds might be some of the strategies to finalize this aim.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Resiquimod, ≥98% (HPLC)