Merck
Attenuation of RNA polymerase II pausing mitigates BRCA1-associated R-loop accumulation and tumorigenesis.
Nature communications (2017-06-27)
Xiaowen Zhang, Huai-Chin Chiang, Yao Wang, Chi Zhang, Sabrina Smith, Xiayan Zhao, Sreejith J Nair, Joel Michalek, Ismail Jatoi, Meeghan Lautner, Boyce Oliver, Howard Wang, Anna Petit, Teresa Soler, Joan Brunet, Francesca Mateo, Miguel Angel Pujana, Elizabeth Poggi, Krysta Chaldekas, Claudine Isaacs, Beth N Peshkin, Oscar Ochoa, Frederic Chedin, Constantine Theoharis, Lu-Zhe Sun, Tyler J Curiel, Richard Elledge, Victor X Jin, Yanfen Hu, Rong Li
ABSTRAKT
Most BRCA1-associated breast tumours are basal-like yet originate from luminal progenitors. BRCA1 is best known for its functions in double-strand break repair and resolution of DNA replication stress. However, it is unclear whether loss of these ubiquitously important functions fully explains the cell lineage-specific tumorigenesis. In vitro studies implicate BRCA1 in elimination of R-loops, DNA-RNA hybrid structures involved in transcription and genetic instability. Here we show that R-loops accumulate preferentially in breast luminal epithelial cells, not in basal epithelial or stromal cells, of BRCA1 mutation carriers. Furthermore, R-loops are enriched at the 5' end of those genes with promoter-proximal RNA polymerase II (Pol II) pausing. Genetic ablation of Cobra1, which encodes a Pol II-pausing and BRCA1-binding protein, ameliorates R-loop accumulation and reduces tumorigenesis in Brca1-knockout mouse mammary epithelium. Our studies show that Pol II pausing is an important contributor to BRCA1-associated R-loop accumulation and breast cancer development.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Cholera Toxin from Vibrio cholerae, ≥90% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Phenol:Chloroform:Isoamyl Alcohol 25:24:1 Saturated with 10 mM Tris, pH 8.0, 1 mM EDTA, Supplied with Equilibration buffer, Molecular Biology
Sigma-Aldrich
Tris-EDTA buffer solution, 100X concentrate for Northern and Southern blotting