Merck
  • Incorporation of cerium oxide into hydroxyapatite coating regulates osteogenic activity of mesenchymal stem cell and macrophage polarization.
Incorporation of cerium oxide into hydroxyapatite coating regulates osteogenic activity of mesenchymal stem cell and macrophage polarization.
Journal of biomaterials applications (2016-12-10)
Kai Li, Qingyi Shen, Youtao Xie, Mingyu You, Liping Huang, Xuebin Zheng
ABSTRAKT
Biomedical coatings for orthopedic implants should facilitate osseointegration and mitigate implant-induced inflammatory reactions. Cerium oxide (CeO
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)