Merck
Function of histone deacetylase 6 as a cofactor of nuclear receptor coregulator LCoR.
The Journal of biological chemistry (2009-09-12)
Ana Palijan, Isabelle Fernandes, Yolande Bastien, Liqun Tang, Mark Verway, Maria Kourelis, Luz E Tavera-Mendoza, Zhi Li, Veronique Bourdeau, Sylvie Mader, Xiang Jiao Yang, John H White
ABSTRAKT
Ligand-dependent corepressor LCoR was identified as a protein that interacts with the estrogen receptor alpha (ERalpha) ligand binding domain in a hormone-dependent manner. LCoR also interacts directly with histone deacetylase 3 (HDAC3) and HDAC6. Notably, HDAC6 has emerged as a marker of breast cancer prognosis. However, although HDAC3 is nuclear, HDAC6 is cytoplasmic in many cells. We found that HDAC6 is partially nuclear in estrogen-responsive MCF7 cells, colocalizes with LCoR, represses transactivation of estrogen-inducible reporter genes, and augments corepression by LCoR. In contrast, no repression was observed upon HDAC6 expression in COS7 cells, where it is exclusively cytoplasmic. LCoR binds to HDAC6 in vitro via a central domain, and repression by LCoR mutants lacking this domain was attenuated. Kinetic chromatin immunoprecipitation assays revealed hormone-dependent recruitment of LCoR to promoters of ERalpha-induced target genes in synchrony with ERalpha. HDAC6 was also recruited to these promoters, and repeat chromatin immunoprecipitation experiments confirmed the corecruitment of LCoR with ERalpha and with HDAC6. Remarkably, however, although we find evidence for corecruitment of LCoR and ERalpha on genes repressed by the receptor, LCoR and HDAC6 failed to coimmunoprecipitate, suggesting that they are part of distinct complexes on these genes. Although small interfering RNA-mediated knockdown of LCoR or HDAC6 augmented expression of an estrogen-sensitive reporter gene in MCF7 cells, unexpectedly their ablation led to reduced expression of some endogenous estrogen target genes. Taken together, these data establish that HDAC6 can function as a cofactor of LCoR but suggest that they may act in enhance expressing some target genes.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
α-Amanitin, from Amanita phalloides, ≥85% (HPLC), powder
Sigma-Aldrich
Przeciwciało anty-HDAC6, CT, Upstate®, from rabbit
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
ANTI-FLAG® M2-Peroxidase (HRP) antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous glycerol solution