Merck
  • Nutritional and Functional Bioactivity Value of Selected Azorean Macroalgae: Ulva compressa, Ulva rigida, Gelidium microdon, and Pterocladiella capillacea.
Nutritional and Functional Bioactivity Value of Selected Azorean Macroalgae: Ulva compressa, Ulva rigida, Gelidium microdon, and Pterocladiella capillacea.
Journal of food science (2017-06-18)
Lisete Paiva, Elisabete Lima, Ana Isabel Neto, Massimo Marcone, José Baptista
ABSTRAKT
This study presents information on the biochemical composition (dry weight basis), nutritional aspects, and angiotensin I-converting enzyme (ACE) inhibitory properties of selected macroalgae (Ulva compressa, Ulva rigida, Gelidium microdon, and Pterocladiella capillacea) from Azores. Moisture content was very high (83.2% to 90.0% of fresh weight). Total dietary fiber (33.7% to 41.0%) that presented a good balance of both soluble and insoluble fibers (15.5% to 19.2% and 18.2% to 21.8%, respectively) was the most abundant component in these macroalgae. Protein and ash (ranged from 15.7% to 23.4% and 10.7% to 20.7%, respectively) were the 2nd most abundant components in red and green macroalgae, respectively. Moderate soluble carbohydrate contents (14.5% to 19.8%) were found in all species. Lipid contents were low (1.0% to 4.3%), particularly in Ulva species (1.0% to 1.7%), but contained higher unsaturated fatty acids (FAs) (7.5% to 32.9% and 29.6% to 69.2% of total FA for monounsaturated fatty acid and polyunsaturated fatty acid groups, respectively) than saturated fatty acid (23.3% to 46.8% of total FA) contents. All the macroalgal proteins had high digestibility in vitro (82.2% to 89.4%, relatively to sodium caseinate), contained high quantity of essential amino acids (45.3% to 58.1% of total amino acids), but in different proportions, and were rich in aspartic and glutamic acids that together account for 17.2% to 36.2% of the total amino acids. These results suggested that regular consumption of the selected macroalgae may improve human health and revealed that they can be used for producing food supplements for human and animal nutrition and/or pharmaceuticals with potential effect on the regional economy. Furthermore, the ACE-inhibitory IC
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trypsin from bovine pancreas, Type I, ~10,000 BAEE units/mg protein
Supelco
Amyloglucosidase solution from Aspergillus niger, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
N-Hexanoyl-L-homoserine lactone, ≥96% (HPLC)
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein
Sigma-Aldrich
Protease from Bacillus licheniformis, lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A