Merck
HMGA2 plays an important role in Cr (VI)-induced autophagy.
International journal of cancer (2017-05-17)
Fan Yang, Lian Zhao, Dan Mei, Liping Jiang, Chengyan Geng, Qiujuan Li, Xiaofeng Yao, Yong Liu, Ying Kong, Jun Cao
ABSTRAKT
Cr (VI) is mutagenic and carcinogenic, but the mechanism is unclear. In this study, the involvement of high mobility group A2 (HMGA2) in Cr (VI)-induced autophagy was investigated. Cr (VI) treatment induced formation of autophagosomes, increased expression of LC3II, Atg12-Atg5, Atg4, Atg10, HMGA1 and HMGA2 proteins, and decreased the expression of p62 in A549 cells. Silencing of HMGA2 gene by siRNA blocked Cr (VI)-induced formation of autophagosomes, expression of LC3II, Atg12-Atg5, Atg10 and reduction of p62. Overexpression of HMGA2 in HEK 293 and HeLa cells could induce the expression of LC3II, Atg12-Atg5 and Atg10, and decrease the expression of p62. Although the protein level of Atg12-Atg5 conjugation changed after Cr (VI) treatment, silencing of HMGA2 and overexpression of HMGA2, both the proteins and mRNA levels of Atg12 and Atg5 were not changed significantly. ChIP assay demonstrated that HMGA2 protein directly bound to the promoter sequence of Atg10 gene, which modulated the conjugation of Atg12-Atg5. Interestingly, 3-MA markedly prevented Cr (VI)-induced cell growth of A549 cells. Our further in vivo study confirmed that the expression of HMGA1, HMGA2, LC3II, Atg12-Atg5, Atg4, Atg5, Atg7, Atg10, Atg12, Beclin 1 were increased and p62 was reduced in lung tissues of Cr (VI)-treated BALB/c mice. Combining, our data demonstrated that HMGA2 plays an important role in Cr (VI)-induced autophagy and the mechanism underlies Atg12-Atg5 conjugation modulated by HMGA2-dependent transcriptional regulation of Atg10. This suggests that HMGA2 might be an important biomarker in Cr (VI)-induced autophagy, cell-growth or other toxicities.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
EZ-Magna ChIP® A - Zestaw do immunoprecypitacji chromatyny, Single day chromatin immunoprecipitation (ChIP) kit containing all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using magnetic A beads. Control primers included.