Merck
  • Probing the Effect of Salinity and pH on Surface Interactions between Air Bubbles and Hydrophobic Solids: Implications for Colloidal Assembly at Air/Water Interfaces.
Probing the Effect of Salinity and pH on Surface Interactions between Air Bubbles and Hydrophobic Solids: Implications for Colloidal Assembly at Air/Water Interfaces.
Chemistry, an Asian journal (2017-04-06)
Xin Cui, Chen Shi, Shuo Zhang, Lei Xie, Jing Liu, Dazhi Jiang, Hongbo Zeng
ABSTRAKT
In this work, a bubble probe atomic force microscope (AFM) was employed to quantify the interactions between two air bubbles and between an air bubble and an octadecyltrichlorosilane (OTS)-hydrophobized mica under various aqueous conditions. The key parameters (e.g., surface potentials, decay length of hydrophobic attraction) were obtained by analyzing the measured forces through a theoretical model based on Reynolds lubrication theory and an augmented Young-Laplace equation by including effect of disjoining pressure. The bubble-OTS hydrophobic attraction with a decay length of 1.0 nm was found to be independent of solution pH and salinity. These parameters were further used to predict the attachment of OTS-hydrophobized particles onto the air/water interface, demonstrating that particle attachment driven by hydrophobic attraction could be facilitated by suppressing electrical double-layer repulsion at low pH or high salinity condition. This facile methodology can be readily extended to quantify interactions of many other colloidal particles with gas/water and oil/water interfaces, with implications for colloidal assembly at different interfaces in many engineering applications.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Trichloro(octadecyl)silane, ≥90%
Sigma-Aldrich
1-Decanethiol, 96%