Merck
Analysis of SDHAF3 in familial and sporadic pheochromocytoma and paraganglioma.
BMC cancer (2017-07-26)
Trisha Dwight, Un Na, Edward Kim, Ying Zhu, Anne Louise Richardson, Bruce G Robinson, Katherine M Tucker, Anthony J Gill, Diana E Benn, Roderick J Clifton-Bligh, Dennis R Winge
ABSTRAKT
Germline mutations in genes encoding subunits of succinate dehydrogenase (SDH) are associated with the development of pheochromocytoma (PC) and/or paraganglioma (PGL). As assembly factors have been identified as playing a role in maturation of individual SDH subunits and assembly of the functioning SDH complex, we hypothesized that SDHAF3 variants may be associated with PC/PGL and functionality of SDH. DNA was extracted from the blood of 37 individuals (from 23 families) with germline SDH mutations and 18 PC/PGL (15 sporadic, 3 familial) and screened for mutations using a custom gene panel, containing SDHAF3 (SDH assembly factor 3) as well as eight known PC/PGL susceptibility genes. Molecular and functional consequences of an identified sequence variant of SDHAF3 were assessed in yeast and mammalian cells (HEK293). Using massively parallel sequencing, we identified a variant in SDHAF3, c.157 T > C (p.Phe53Leu), associated with increased prevalence in familial and sporadic PC/PGL (6.6%) when compared to normal populations (1.2% [1000 Genomes], p = 0.003; 2.1% [Exome Aggregation Consortium], p = 0.0063). In silico prediction tools suggest this variant is probably damaging to protein function, hence we assessed molecular and functional consequences of the resulting amino acid change (p.Phe53Leu) in yeast and human cells. We showed that introduction of SDHAF3 p.Phe53Leu into Sdh7 (ortholog of SDHAF3 in humans) null yeast resulted in impaired function, as observed by its failure to restore SDH activity when expressed in Sdh7 null yeast relative to WT SDHAF3. As SDHAF3 is involved in maturation of SDHB, we tested the functional impact of SDHAF3 c.157 T > C and various clinically relevant SDHB mutations on this interaction. Our in vitro studies in human cells show that SDHAF3 interacts with SDHB (residues 46 and 242), with impaired interaction observed in the presence of the SDHAF3 c.157 T > C variant. Our studies reveal novel insights into the biogenesis of SDH, uncovering a vital interaction between SDHAF3 and SDHB. We have shown that SDHAF3 interacts directly with SDHB (residue 242 being key to this interaction), and that a variant in SDHAF3 (c.157 T > C [p.Phe53Leu]) may be more prevalent in individuals with PC/PGL, and is hypomorphic via impaired interaction with SDHB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Roche
Anty-GFP, from mouse IgG1κ (clones 7.1 and 13.1)