Merck
Oncolytic Adenovirus and Tumor-Targeting Immune Modulatory Therapy Improve Autologous Cancer Vaccination.
Cancer research (2017-06-02)
Hong Jiang, Yisel Rivera-Molina, Candelaria Gomez-Manzano, Karen Clise-Dwyer, Laura Bover, Luis M Vence, Ying Yuan, Frederick F Lang, Carlo Toniatti, Mohammad B Hossain, Juan Fueyo
ABSTRAKT
Oncolytic viruses selectively lyse tumor cells, disrupt immunosuppression within the tumor, and reactivate antitumor immunity, but they have yet to live up to their therapeutic potential. Immune checkpoint modulation has been efficacious in a variety of cancer with an immunogenic microenvironment, but is associated with toxicity due to nonspecific T-cell activation. Therefore, combining these two strategies would likely result in both effective and specific cancer therapy. To test the hypothesis, we first constructed oncolytic adenovirus Delta-24-RGDOX expressing the immune costimulator OX40 ligand (OX40L). Like its predecessor Delta-24-RGD, Delta-24-RGDOX induced immunogenic cell death and recruit lymphocytes to the tumor site. Compared with Delta-24-RGD, Delta-24-RGDOX exhibited superior tumor-specific activation of lymphocytes and proliferation of CD8
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Ethidium homodimer, suitable for fluorescence, ~90% (HPCE)