- Alzheimer's disease markers in the aged sheep (Ovis aries).
Alzheimer's disease markers in the aged sheep (Ovis aries).
Neurobiology of aging (2017-07-21)
Suzanne J Reid, Natasha E Mckean, Kristen Henty, Erik Portelius, Kaj Blennow, Skye R Rudiger, C Simon Bawden, Renee R Handley, Paul J Verma, Richard L M Faull, Henry J Waldvogel, Henrik Zetterberg, Russell G Snell
PMID28728117
ABSTRAKT
This study reports the identification and characterization of markers of Alzheimer's disease (AD) in aged sheep (Ovis aries) as a preliminary step toward making a genetically modified large animal model of AD. Importantly, the sequences of key proteins involved in AD pathogenesis are highly conserved between sheep and human. The processing of the amyloid-β (Aβ) protein is conserved between sheep and human, and sheep Aβ
MATERIAŁY