Merck
  • Gamma-tocotrienol and gamma-tocopherol are primarily metabolized to conjugated 2-(beta-carboxyethyl)-6-hydroxy-2,7,8-trimethylchroman and sulfated long-chain carboxychromanols in rats.
Gamma-tocotrienol and gamma-tocopherol are primarily metabolized to conjugated 2-(beta-carboxyethyl)-6-hydroxy-2,7,8-trimethylchroman and sulfated long-chain carboxychromanols in rats.
The Journal of nutrition (2009-03-20)
Helene Freiser, Qing Jiang
ABSTRAKT
The metabolism of gamma-tocotrienol (gamma-TE) and gamma-tocopherol (gamma-T) was investigated in human A549 cells and in rats. Similar to gamma-T, A549 cells metabolized gamma-TE to sulfated 9'-, 11'-, and 13'-carboxychromanol and their unconjugated counterparts. After 72-h incubation with the cells, 90% of long-chain carboxychromanols in the culture media from gamma-TE, but <45% from gamma-T, were in the sulfated form. The formation of these metabolites was further investigated in rats gavaged by gamma-TE at 10 or 50 mg/kg, gamma-T at 10 mg/kg, or tocopherol-stripped corn oil in controls. Six hours after a single dosing, the supplemented rats had increased plasma concentrations of 13'-carboxychromanol and sulfated 9'-, 11'-, 13'-carboxychromanol, whereas none of these metabolites were detectable in the controls. Sulfated 11'-carboxychromanol was the most abundant long-chain metabolite in gamma-TE-supplemented rats. Sulfatase/glucuronidase hydrolysis revealed for the first time that >88% 2-(beta-carboxyethyl)-6-hydroxychroman (gamma-CEHC), the terminal beta-oxidation metabolite, was in the conjugated form in the plasma. In all groups, conjugated gamma-CEHC accounted for >75% of total metabolites, whereas free CEHC was a minor metabolite. At 10 mg/kg, the plasma concentrations of total metabolites from gamma-TE-supplemented rats were higher (P < 0.05) than those from gamma-T-fed rats. These results demonstrate that in rats, conjugation such as sulfation occurs parallel to beta-oxidation in the liver and is quantitatively important to vitamin E metabolism. Conjugated long-chain carboxychromanols may be novel excreted metabolites during supplementation. Our data also provide in vivo evidence that gamma-TE is more extensively metabolized than gamma-T.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid