- SOX11 promotes tumor protective microenvironment interactions through CXCR4 and FAK regulation in mantle cell lymphoma.
SOX11 promotes tumor protective microenvironment interactions through CXCR4 and FAK regulation in mantle cell lymphoma.
Blood (2017-05-24)
Patricia Balsas, Jara Palomero, Álvaro Eguileor, Marta Leonor Rodríguez, Maria Carmela Vegliante, Ester Planas-Rigol, Marta Sureda-Gómez, Maria C Cid, Elias Campo, Virginia Amador
PMID28533307
ABSTRAKT
SOX11 overexpression in mantle cell lymphoma (MCL) has been associated with more aggressive behavior and worse outcome. However, SOX11 oncogenic pathways driving MCL tumor progression are poorly understood. Here, we demonstrate that SOX11 binds to regulatory regions of 2 important genes for microenvironment signals in cancer: (C-X-C motif) chemokine receptor 4 (