Merck
  • 3D-printable, highly conductive hybrid composites employing chemically-reinforced, complex dimensional fillers and thermoplastic triblock copolymers.
3D-printable, highly conductive hybrid composites employing chemically-reinforced, complex dimensional fillers and thermoplastic triblock copolymers.
Nanoscale (2017-02-10)
Yejin Jo, Ju Young Kim, So-Yun Kim, Yeong-Hui Seo, Kwang-Suk Jang, Su Yeon Lee, Sungmook Jung, Beyong-Hwan Ryu, Hyun-Suk Kim, Jang-Ung Park, Youngmin Choi, Sunho Jeong
ABSTRAKT
The use of 3-dimensional (3D) printable conductive materials has gained significant attention for various applications because of their ability to form unconventional geometrical architectures that cannot be realized with traditional 2-dimensional printing techniques. To resolve the major requisites in printed electrodes for practical applications (including high conductivity, 3D printability, excellent adhesion, and low-temperature processability), we have designed a chemically-reinforced multi-dimensional filler system comprising amine-functionalized carbon nanotubes, carboxyl-terminated silver nanoparticles, and Ag flakes, with the incorporation of a thermoplastic polystyrene-polyisoprene-polystyrene (SIS) triblock copolymer. It is demonstrated that both high conductivity, 22 939 S cm
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DCB, ≥98% (HPLC), solid
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Octylamine, 99%
Sigma-Aldrich
PTCDA, 97%
Sigma-Aldrich
Phenylhydrazine, 97%
Sigma-Aldrich
Oleic acid, technical grade, 90%