Merck
Teuvincenone F Suppresses LPS-Induced Inflammation and NLRP3 Inflammasome Activation by Attenuating NEMO Ubiquitination.
Frontiers in pharmacology (2017-09-08)
Xibao Zhao, Debing Pu, Zizhao Zhao, Huihui Zhu, Hongrui Li, Yaping Shen, Xingjie Zhang, Ruihan Zhang, Jianzhong Shen, Weilie Xiao, Weilin Chen
ABSTRAKT
Inflammation causes many diseases that are serious threats to human health. However, the molecular mechanisms underlying regulation of inflammation and inflammasome activation are not fully understood which has delayed the discovery of new anti-inflammatory drugs of urgent clinic need. Here, we found that the natural compound Teuvincenone F, which was isolated and purified from the stems and leaves of
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli O111:B4, purified by ion-exchange chromatography, TLR ligand tested
Sigma-Aldrich
PR-619, ≥95% (HPLC)