Merck
α-Synuclein Amyloids Hijack Prion Protein to Gain Cell Entry, Facilitate Cell-to-Cell Spreading and Block Prion Replication.
Scientific reports (2017-09-01)
Suzana Aulić, Lara Masperone, Joanna Narkiewicz, Elisa Isopi, Edoardo Bistaffa, Elena Ambrosetti, Beatrice Pastore, Elena De Cecco, Denis Scaini, Paola Zago, Fabio Moda, Fabrizio Tagliavini, Giuseppe Legname
ABSTRAKT
The precise molecular mechanism of how misfolded α-synuclein (α-Syn) accumulates and spreads in synucleinopathies is still unknown. Here, we show the role of the cellular prion protein (PrP
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture